siRNA Details
| Gene Symbol: Klrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010654.3 | NP_034784.1 | 6 | 602 | ||
| GenBank mRNA | AF030311.1 | - | 5 | 582 | ||
| AF030312.1 | - | 6 | 631 | |||
| AF057714.1 | - | 5 | 582 | |||
| AK136548.1 | - | 6 | 603 | |||
| AK162976.1 | - | 7 | 663 | |||

Klrd1
Genomic Map