siRNA Details
| Gene Symbol: Krtap13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010671.1 | NP_034801.1 | 1 | 538 | ||
| GenBank mRNA | AF031485.1 | - | 1 | 538 | ||
| AK003960.1 | - | 1 | 561 | |||
| BC063754.1 | - | 1 | 545 | 30293948 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap13| Gene Symbol: Krtap13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010671.1 | NP_034801.1 | 1 | 538 | ||
| GenBank mRNA | AF031485.1 | - | 1 | 538 | ||
| AK003960.1 | - | 1 | 561 | |||
| BC063754.1 | - | 1 | 545 | 30293948 | ||