siRNA Details
| Gene Symbol: Laptm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010686.3 | NP_034816.1 | 8 | 2221 | ||
| GenBank mRNA | AK031802.1 | - | 8 | 2233 | ||
| AK089508.1 | - | 9 | 2172 | |||
| AK089535.1 | - | 9 | 2164 | |||
| AK150299.1 | - | 9 | 2199 | |||
| AK152918.1 | - | 9 | 1836 | |||
| AK153877.1 | - | 8 | 2235 | |||
| AK155353.1 | - | 3 | 1678 | |||
| AK155744.1 | - | 9 | 2053 | |||
| AK163146.1 | - | 8 | 2236 | |||
| AK170690.1 | - | 10 | 2028 | |||
| AK170867.1 | - | 9 | 1915 | |||
| AK171053.1 | - | 9 | 2172 | |||
| AK171057.1 | - | 8 | 2272 | |||
| AK171116.1 | - | 9 | 2164 | |||
| AK171193.1 | - | 9 | 2164 | |||
| AK171213.1 | - | 8 | 2236 | |||
| AK171236.1 | - | 9 | 1947 | |||
| AK171244.1 | - | 8 | 2236 | |||
| AK171250.1 | - | 9 | 2005 | |||
| AK171269.1 | - | 8 | 2273 | |||
| AK171270.1 | - | 8 | 2239 | |||
| AK171274.1 | - | 8 | 2229 | |||
| AK171280.1 | - | 8 | 2269 | |||
| AK171285.1 | - | 8 | 2273 | |||
| AK171481.1 | - | 8 | 2309 | |||
| AK171547.1 | - | 9 | 2133 | |||
| AK171562.1 | - | 9 | 2158 | |||
| AK171571.1 | - | 9 | 2184 | |||
| AK171575.1 | - | 9 | 2202 | |||
| AK172306.1 | - | 9 | 1880 | |||
| AK172486.1 | - | 8 | 2271 | |||
| AK172707.1 | - | 9 | 1948 | |||
| AK172717.1 | - | 6 | 1870 | |||
| BC020993.1 | - | 8 | 2293 | 4037686 | ||
| BC055057.1 | - | 8 | 2280 | 6529760 | ||
| U29539.1 | - | 8 | 2277 | |||

Laptm5
Genomic Map