siRNA Details
| Gene Symbol: Il1rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025602.3 | NP_001020773.1 | 3 | 466 | ||
| NM_001294171.1 | NP_001281100.1 | 3 | 299 | |||
| NM_010743.3 | NP_034873.2 | 3 | 466 | |||
| GenBank mRNA | AK033417.1 | - | 3 | 319 | ||
| AK145118.1 | - | 3 | 320 | |||
| BC051445.1 | - | 3 | 276 | 30073421 | ||
| BY228016.1 | - | 3 | 300 | |||
| D13695.1 | - | 2 | 158 | |||
| M24843.1 | - | 3 | 307 | |||
| Y07519.1 | - | 3 | 310 | |||

Il1rl1
Genomic Map