siRNA Details
| Gene Symbol: Mcpt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010779.2 | NP_034909.2 | 4 | 529 | ||
| GenBank mRNA | AB051900.1 | - | 4 | 477 | ||
| AK144385.1 | - | 4 | 530 | |||
| AY007569.1 | - | 4 | 529 | |||
| M55617.1 | - | 529 | ||||
| X68804.1 | - | 4 | 476 | |||

Mcpt4
Genomic Map