siRNA Details
| Gene Symbol: Mid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290504.1 | NP_001277433.1 | 479 | |||
| NM_001290505.1 | NP_001277434.1 | 703 | ||||
| NM_001290506.1 | NP_001277435.1 | 703 | ||||
| NM_010797.3 | NP_034927.2 | 586 | ||||
| GenBank mRNA | AF026565.1 | - | 479 | |||
| AK135734.1 | - | 702 | ||||
| BC053704.1 | - | 670 | 30058736 | |||
| Y14848.1 | - | 540 | ||||

Mid1
Genomic Map