siRNA Details
| Gene Symbol: Marcksl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010807.4 | NP_034937.1 | 1, 2 | 332 | ||
| GenBank mRNA | AK079390.1 | - | 1, 2 | 281 | ||
| AK083913.1 | - | 1, 2 | 283 | |||
| AK150261.1 | - | 1, 2 | 280 | |||
| AK152990.1 | - | 1, 2 | 280 | |||
| AK169355.1 | - | 1, 2 | 283 | |||
| BC006757.1 | - | 1, 2 | 260 | 3257582 | ||
| CX243729.1 | - | 1, 2 | 332 | |||
| X61399.1 | - | 1, 2 | 269 | |||

Marcksl1
Genomic Map