siRNA Details
| Gene Symbol: Magoh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282737.1 | NP_001269666.1 | 2, 3 | 258 | ||
| GenBank mRNA | AF007862.1 | - | 2, 3 | 245 | ||
| AF035939.1 | - | 2, 3 | 205 | |||
| AK011512.1 | - | 2, 3 | 197 | |||
| BC018176.1 | - | 2, 3 | 196 | 3587774 | ||
| BY134232.1 | - | 2, 3 | 259 | |||

Magoh
Genomic Map