siRNA Details
| Gene Symbol: Mal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171187.1 | NP_001164658.1 | 3 | 1061 | ||
| NM_010762.5 | NP_034892.1 | 4 | 1229 | |||
| GenBank mRNA | AK004639.1 | - | 4 | 816 | ||
| AK158653.1 | - | 4 | 1230 | |||
| AK169172.1 | - | 4 | 753 | |||
| BC006826.1 | - | 4 | 726 | 3597561 | ||
| Y07626.1 | - | 4 | 705 | |||

Mal
Genomic Map