siRNA Details
| Gene Symbol: Mbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010773.2 | NP_034903.2 | 1 | 628 | ||
| GenBank mRNA | AF072243.1 | - | 1 | 586 | ||
| AF072245.1 | - | 1 | 586 | |||
| AK082825.1 | - | 1 | 65 | |||
| AK147008.1 | - | 1 | 513 | |||
| BC026698.1 | - | 1 | 588 | |||
| BC046607.1 | - | 1 | 628 | 6399206 | ||
| BC057289.1 | - | 1 | 460 | |||
| BC058864.1 | - | 1 | 448 | |||

Mbd2
Genomic Map