siRNA Details
| Gene Symbol: Mdk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012335.2 | NP_001012335.1 | 5 | 584 | ||
| NM_001012336.2 | NP_001012336.1 | 5 | 581 | |||
| NM_001291481.1 | NP_001278410.1 | 5 | 491 | |||
| NM_001291482.1 | NP_001278411.1 | 4 | 504 | |||
| NM_001291483.1 | NP_001278412.1 | 4 | 621 | |||
| NM_010784.5 | NP_034914.1 | 5 | 783 | |||
| GenBank mRNA | AK032742.1 | - | 5 | 487 | ||
| AK084457.1 | - | 5 | 504 | |||
| AY864926.1 | - | 5 | 517 | |||
| BC012244.1 | - | 5 | 452 | 4167496 | ||
| CF728526.1 | - | 5 | 477 | |||
| CF738400.1 | - | 5 | 589 | |||
| CF740555.1 | - | 3 | 301 | |||
| CJ174578.1 | - | 4 | 379 | |||
| DQ323886.1 | - | 5 | 559 | |||
| DQ323887.1 | - | 4 | 592 | |||
| M19662.1 | - | 4 | 662 | |||
| M34327.1 | - | 5 | 457 | |||
| M34328.1 | - | 5 | 768 | |||
| M35833.1 | - | 5 | 457 | |||

Mdk
Genomic Map