siRNA Details
| Gene Symbol: Limk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139231.1 | - | 2 | 2637 | ||
| AK141813.1 | - | 9 | 1292 | |||
| AK170166.1 | - | 8 | 1067 | |||
| AK172083.1 | - | 8 | 1051 | |||
| BC063768.1 | - | 9 | 1272 | |||
| U14166.1 | - | 9 | 1315 | |||
| U15159.1 | - | 9 | 1287 | |||

Limk1
Genomic Map