siRNA Details
| Gene Symbol: M6pr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010749.6 | NP_034879.2 | 7 | 2138 | ||
| GenBank mRNA | AK133943.1 | - | 7 | 2144 | ||
| AK149177.1 | - | 7 | 2168 | |||
| AK151607.1 | - | 7 | 2199 | |||
| AK153140.1 | - | 7 | 2149 | |||
| AK153302.1 | - | 7 | 2219 | |||
| AK153474.1 | - | 7 | 2159 | |||
| BC027210.1 | - | 7 | 2094 | |||
| BC046956.1 | - | 7 | 2138 | |||
| BC080811.1 | - | 7 | 2138 | 6438831 | ||
| M63286.1 | - | 7 | 2046 | |||
| X64068.1 | - | 7 | 2140 | |||

M6pr
Genomic Map