siRNA Details
| Gene Symbol: Melk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010790.2 | NP_034920.2 | 17, 18 | 1955 | ||
| GenBank mRNA | AK011932.1 | - | 17, 18 | 1955 | ||
| AK129076.1 | - | 17, 18 | 1908 | |||
| AK137000.1 | - | 17, 18 | 1982 | |||
| AK145316.1 | - | 17, 18 | 1864 | |||
| AK161029.1 | - | 17, 18 | 1964 | |||
| AK164138.1 | - | 17, 18 | 1922 | |||
| AK167457.1 | - | 17, 18 | 1879 | |||
| AK206998.1 | - | 1, 2 | 55 | |||
| BC085276.1 | - | 17, 18 | 1988 | 30443236 | ||
| L76158.1 | - | 17, 18 | 1886 | |||
| X95351.1 | - | 17, 18 | 1893 | |||

Melk
Genomic Map