siRNA Details
| Gene Symbol: Marcksl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010807.4 | NP_034937.1 | 2 | 1583 | ||
| GenBank mRNA | AK150261.1 | - | 2 | 1530 | ||
| AK152990.1 | - | 2 | 1531 | |||
| AK169355.1 | - | 2 | 1534 | |||
| BC006757.1 | - | 2 | 1509 | 3257582 | ||
| X61399.1 | - | 2 | 1521 | |||

Marcksl1
Genomic Map