siRNA Details
| Gene Symbol: GLIPR1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304964.1 | NP_001291893.1 | 4 | 654 | ||
| NM_152779.3 | NP_689992.1 | 4 | 654 | |||
| XM_011538134.2 | XP_011536436.2 | 4 | 3449 | |||
| XM_017019134.1 | XP_016874623.1 | 2 | 3112 | |||
| GenBank mRNA | AY358731.1 | - | 4 | 654 | ||
| BC014603.1 | - | 4 | 615 | 4822995 | ||

GLIPR1L1
Genomic Map