siRNA Details
| Gene Symbol: NFXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278623.1 | NP_001265552.1 | 21 | 2519 | ||
| NM_001278624.1 | NP_001265553.1 | 21 | 2657 | |||
| NM_152995.5 | NP_694540.3 | 21 | 2559 | |||
| NR_103795.1 | 22 | 2699 | ||||
| GenBank mRNA | AB085695.1 | - | 21 | 2515 | ||
| AB181916.1 | - | 21 | 2639 | |||
| AK131285.1 | - | 19 | 2105 | |||
| AK302440.1 | - | 21 | 2517 | |||
| AY134856.1 | - | 5 | 563 | |||
| BC018019.1 | - | 4 | 254 | |||
| BC051193.1 | - | 22 | 2670 | 5271401 | ||

NFXL1
Genomic Map