siRNA Details
| Gene Symbol: Nfe2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010902.3 | NP_035032.1 | 2 | 315 | ||
| GenBank mRNA | AK142347.1 | - | 2 | 217 | ||
| AK144247.1 | - | 2 | 189 | |||
| BC003340.1 | - | 2 | 171 | |||
| BC026943.1 | - | 2 | 256 | 3663276 | ||
| BY792550.1 | - | 2 | 315 | |||
| U20532.1 | - | 2 | 331 | |||

Nfe2l2
Genomic Map