siRNA Details
| Gene Symbol: Map6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048167.1 | NP_001041632.1 | 2, 3 | 2077 | ||
| NM_010837.3 | NP_034967.2 | 2, 3 | 2077 | |||
| GenBank mRNA | AK018193.2 | - | 2, 3 | 1557 | ||
| AK087413.1 | - | 2, 3 | 2040 | |||
| BC053039.1 | - | 2, 3 | 2078 | 5709455 | ||

Map6
Genomic Map