siRNA Details
| Gene Symbol: Nfkbib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK010218.1 | - | 3 | 652 | ||
| AK038631.1 | - | 3 | 700 | |||
| AK156731.1 | - | 3 | 626 | |||
| AY176058.1 | - | 3 | 659 | |||
| AY176059.1 | - | 3 | 690 | |||
| BC021938.1 | - | 3 | 657 | 5136809 | ||
| U19799.1 | - | 3 | 671 | |||

Nfkbib
Genomic Map