siRNA Details
| Gene Symbol: Msn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010833.2 | NP_034963.2 | 7 | 932 | ||
| GenBank mRNA | AK088336.1 | - | 7 | 880 | ||
| AK133819.1 | - | 7 | 936 | |||
| AK145746.1 | - | 7 | 936 | |||
| AK150869.1 | - | 7 | 935 | |||
| AK153210.1 | - | 7 | 935 | |||
| AK157701.1 | - | 7 | 887 | |||
| AK216494.1 | - | 2 | 208 | |||
| BC003474.1 | - | 7 | 923 | |||
| BC047366.1 | - | 7 | 926 | 3711212 | ||
| BQ950368.1 | - | 1 | 69 | |||
| CF584663.1 | - | 420 | ||||
| M86390.1 | - | 6 | 712 | |||
| S47577.1 | - | 7 | 917 | |||

Msn
Genomic Map