siRNA Details
| Gene Symbol: Myo15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103171.1 | NP_001096641.1 | 10, 11 | 4455 | ||
| NM_010862.2 | NP_034992.2 | 10, 11 | 4455 | |||
| NM_182698.2 | NP_874357.2 | 9, 10 | 693 | |||
| GenBank mRNA | AB014510.1 | - | 6, 7 | 458 | ||
| AF053130.1 | - | 8, 9 | 661 | |||
| AF144095.1 | - | 10, 11 | 4455 | |||
| AY331132.1 | - | 10, 11 | 4455 | |||
| AY331133.1 | - | 9, 10 | 693 | |||

Myo15
Genomic Map