siRNA Details
| Gene Symbol: Nnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010924.2 | NP_035054.1 | 3 | 538 | ||
| GenBank mRNA | AK006371.1 | - | 2 | 285 | ||
| BC028757.1 | - | 3 | 454 | 1067421 | ||
| U86105.1 | - | 3 | 585 | |||
| U86106.1 | - | 3 | 585 | |||
| U86107.1 | - | 3 | 585 | |||
| U86108.1 | - | 3 | 585 | |||

Nnmt
Genomic Map