siRNA Details
| Gene Symbol: Uhrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111078.1 | NP_001104548.1 | 2 | 187 | ||
| NM_001111079.1 | NP_001104549.1 | 2 | 331 | |||
| NM_001111080.1 | NP_001104550.1 | 2 | 187 | |||
| NM_010931.3 | NP_035061.3 | 2 | 331 | |||
| GenBank mRNA | AF274046.1 | - | 2 | 244 | ||
| AK143688.1 | - | 2 | 156 | |||
| AK145376.1 | - | 2 | 153 | |||
| AK145419.1 | - | 2 | 152 | |||
| AK145543.1 | - | 2 | 271 | |||
| AK145583.1 | - | 2 | 153 | |||
| AK146951.1 | - | 2 | 304 | |||
| AK147046.1 | - | 2 | 271 | |||
| AK150489.1 | - | 2 | 187 | |||
| AK151701.1 | - | 2 | 153 | |||
| AK151837.1 | - | 2 | 187 | |||
| AK152930.1 | - | 2 | 187 | |||
| AK153083.1 | - | 2 | 187 | |||
| BC022167.1 | - | 2 | 259 | 4017286 | ||
| D87908.1 | - | 2 | 242 | |||

Uhrf1
Genomic Map