siRNA Details
| Gene Symbol: Olfr54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010997.1 | NP_035127.1 | 1 | 283 | ||
| GenBank mRNA | AF102530.1 | - | 1 | 88 | ||
| BC099884.1 | - | 1 | 282 | |||
| BC099885.1 | - | 1 | 282 | |||
| BC100678.1 | - | 1 | 282 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr54| Gene Symbol: Olfr54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010997.1 | NP_035127.1 | 1 | 283 | ||
| GenBank mRNA | AF102530.1 | - | 1 | 88 | ||
| BC099884.1 | - | 1 | 282 | |||
| BC099885.1 | - | 1 | 282 | |||
| BC100678.1 | - | 1 | 282 | |||