siRNA Details
| Gene Symbol: IER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003897.3 | NP_003888.2 | 2 | 882 | ||
| GenBank mRNA | AF039067.1 | - | 1 | 959 | ||
| AK222588.1 | - | 2 | 879 | |||
| BC000844.2 | - | 2 | 866 | 3457670 | ||
| BC005080.1 | - | 2 | 868 | 3617088 | ||
| S81914.1 | - | 2 | 865 | |||
| Y14551.1 | - | 2 | 861 | |||

IER3
Genomic Map