siRNA Details
| Gene Symbol: Pdpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF086625.1 | - | 2, 3 | 388 | ||
| AY062008.1 | - | 2, 3 | 318 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdpk1| Gene Symbol: Pdpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF086625.1 | - | 2, 3 | 388 | ||
| AY062008.1 | - | 2, 3 | 318 | |||