siRNA Details
| Gene Symbol: Phxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC119608.2 | - | 1 | 179 | 40061632 | |
| BC119609.2 | - | 1 | 179 | 40061635 | ||
| X12809.1 | - | 1 | 179 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phxr1| Gene Symbol: Phxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC119608.2 | - | 1 | 179 | 40061632 | |
| BC119609.2 | - | 1 | 179 | 40061635 | ||
| X12809.1 | - | 1 | 179 | |||