siRNA Details
| Gene Symbol: Pik3c2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011084.2 | NP_035214.2 | 2 | 339 | ||
| NM_207683.2 | NP_997566.1 | 3085 | ||||
| GenBank mRNA | AB008791.1 | - | 3085 | |||
| AB008792.1 | - | 2 | 334 | |||
| AK036916.1 | - | 2 | 150 | |||
| AK054266.1 | - | 2 | 130 | |||
| AK054271.1 | - | 2 | 215 | |||
| AK054415.1 | - | 2 | 282 | |||
| AK076317.1 | - | 2740 | ||||
| BC061142.1 | - | 2 | 126 | |||
| BC150813.1 | - | 3077 | 9087724 | |||

Pik3c2g
Genomic Map