siRNA Details
| Gene Symbol: Pira1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011087.1 | NP_035217.1 | 1798 | |||
| GenBank mRNA | U96682.1 | - | 1798 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pira1| Gene Symbol: Pira1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011087.1 | NP_035217.1 | 1798 | |||
| GenBank mRNA | U96682.1 | - | 1798 | |||