siRNA Details
| Gene Symbol: GPR101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054021.1 | NP_473362.1 | 1 | 971 | ||
| GenBank mRNA | BC069439.1 | - | 1 | 971 | 7262356 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR101| Gene Symbol: GPR101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054021.1 | NP_473362.1 | 1 | 971 | ||
| GenBank mRNA | BC069439.1 | - | 1 | 971 | 7262356 | |
Set Membership: |
|