siRNA Details
| Gene Symbol: Prkce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011104.3 | NP_035234.1 | 2 | 1183 | ||
| GenBank mRNA | AF028009.1 | - | 2 | 384 | ||
| AF325507.1 | - | 2 | 375 | |||
| AK082438.1 | - | 2 | 1184 | |||
| AK085351.1 | - | 3 | 595 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkce| Gene Symbol: Prkce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011104.3 | NP_035234.1 | 2 | 1183 | ||
| GenBank mRNA | AF028009.1 | - | 2 | 384 | ||
| AF325507.1 | - | 2 | 375 | |||
| AK082438.1 | - | 2 | 1184 | |||
| AK085351.1 | - | 3 | 595 | |||