siRNA Details
| Gene Symbol: Pa2g4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011119.3 | NP_035249.1 | 2 | 269 | ||
| GenBank mRNA | AK002685.1 | - | 2 | 347 | ||
| AK076286.1 | - | 2 | 274 | |||
| AK144645.1 | - | 2 | 323 | |||
| AK166936.1 | - | 2 | 278 | |||
| AK168582.1 | - | 2 | 271 | |||
| BC003777.1 | - | 2 | 259 | |||
| BC005593.1 | - | 2 | 259 | |||
| BC028926.1 | - | 2 | 252 | |||
| BC037623.1 | - | 2 | 289 | |||
| BC046532.1 | - | 2 | 223 | 5358060 | ||
| BC086788.1 | - | 2 | 2139 | |||
| U43918.1 | - | 2 | 428 | |||
| X84789.1 | - | 2 | 334 | |||

Pa2g4
Genomic Map