siRNA Details
| Gene Symbol: Pex11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011068.1 | NP_035198.1 | 3 | 820 | ||
| GenBank mRNA | AF093669.1 | - | 3 | 820 | ||
| AK076776.1 | - | 2 | 810 | |||
| AK078954.1 | - | 2 | 653 | |||
| AK149387.1 | - | 3 | 819 | |||
| BC023439.1 | - | 3 | 793 | 5251140 | ||
| BC028786.1 | - | 1 | 1402 | |||

Pex11a
Genomic Map