siRNA Details
| Gene Symbol: Prkca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011101.3 | NP_035231.2 | 1 | 328 | ||
| GenBank mRNA | AK147084.1 | - | 1 | 329 | ||
| BC096493.1 | - | 1 | 122 | 30742571 | ||
| M25811.1 | - | 1 | 362 | |||
| X52684.1 | - | 2 | 269 | |||
| X52685.1 | - | 2 | 269 | |||

Prkca
Genomic Map