siRNA Details
| Gene Symbol: Pcbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103165.1 | NP_001096635.1 | 6, 7 | 616 | ||
| NM_001103166.1 | NP_001096636.1 | 6, 7 | 616 | |||
| NM_001174073.1 | NP_001167544.1 | 6, 7 | 592 | |||
| NM_011042.2 | NP_035172.2 | 6, 7 | 616 | |||
| GenBank mRNA | AK012297.1 | - | 6, 7 | 464 | ||
| AK161527.1 | - | 6, 7 | 440 | |||
| AK169268.1 | - | 6, 7 | 710 | |||
| BC020478.1 | - | 5, 6 | 361 | |||
| BQ900329.1 | - | 6, 7 | 592 | |||
| EU604548.1 | - | 5, 6 | 371 | |||
| L19661.1 | - | 6, 7 | 503 | |||

Pcbp2
Genomic Map