siRNA Details
| Gene Symbol: Pdgfra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083316.1 | NP_001076785.1 | 23 | 4877 | ||
| NM_011058.2 | NP_035188.2 | 23 | 4860 | |||
| GenBank mRNA | AK147267.1 | - | 23 | 4809 | ||
| M84607.1 | - | 23 | 4867 | |||

Pdgfra
Genomic Map