siRNA Details
| Gene Symbol: Pdpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080773.2 | NP_001074242.1 | 11 | 1458 | ||
| NM_001286662.1 | NP_001273591.1 | 11 | 1376 | |||
| NM_011062.4 | NP_035192.2 | 11 | 1458 | |||
| GenBank mRNA | AF079535.1 | - | 11 | 1415 | ||
| AF086625.1 | - | 11 | 1424 | |||
| AF126294.1 | - | 11 | 1399 | |||
| AK147149.1 | - | 11 | 1450 | |||
| AK147835.1 | - | 11 | 1433 | |||
| AK149162.1 | - | 11 | 1392 | |||
| AK162677.1 | - | 11 | 1418 | |||
| AK165564.1 | - | 11 | 1396 | |||
| AK188106.1 | - | 3 | 426 | |||
| AK205439.1 | - | 2 | 371 | |||
| AK215448.1 | - | 2 | 247 | |||
| AK219503.1 | - | 3 | 385 | |||
| AY062008.1 | - | 11 | 1354 | |||
| BC146001.1 | - | 11 | 1395 | 40131304 | ||
| BC146003.1 | - | 11 | 1395 | 40131306 | ||

Pdpk1
Genomic Map