siRNA Details
| Gene Symbol: Prkca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC096493.1 | - | 17 | 2126 | 30742571 | |
| M25811.1 | - | 17 | 2364 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkca| Gene Symbol: Prkca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC096493.1 | - | 17 | 2126 | 30742571 | |
| M25811.1 | - | 17 | 2364 | |||