siRNA Details
| Gene Symbol: Prkce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011104.3 | NP_035234.1 | 8, 9 | 1878 | ||
| GenBank mRNA | AF028009.1 | - | 8, 9 | 1079 | ||
| AF325507.1 | - | 8, 9 | 1070 | |||
| AK082438.1 | - | 8, 9 | 1879 | |||
| AK085351.1 | - | 9, 10 | 1290 | |||

Prkce
Genomic Map