siRNA Details
| Gene Symbol: Ppara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113418.1 | NP_001106889.1 | 3 | 344 | ||
| NM_011144.6 | NP_035274.2 | 3 | 582 | |||
| GenBank mRNA | AK035676.1 | - | 3 | 583 | ||
| AK081709.1 | - | 3 | 345 | |||
| AK149460.1 | - | 3 | 281 | |||
| BC016892.1 | - | 3 | 575 | 4222384 | ||
| X57638.1 | - | 3 | 303 | |||

Ppara
Genomic Map