siRNA Details
| Gene Symbol: Psen2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128605.1 | NP_001122077.1 | 3 | 239 | ||
| NM_011183.3 | NP_035313.2 | 3 | 248 | |||
| GenBank mRNA | AF038935.1 | - | 2 | 176 | ||
| AK014706.1 | - | 2 | 183 | |||
| AK148111.1 | - | 3 | 240 | |||
| AK154119.1 | - | 3 | 184 | |||
| BC010403.1 | - | 2 | 194 | 4038761 | ||
| BY136632.1 | - | 3 | 248 | |||
| U57324.1 | - | 3 | 199 | |||

Psen2
Genomic Map