siRNA Details
| Gene Symbol: Psma3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011184.4 | NP_035314.3 | 2 | 219 | ||
| GenBank mRNA | AF055983.1 | - | 2 | 137 | ||
| AF060089.1 | - | 2 | 105 | |||
| AI303320.1 | - | 2 | 218 | |||
| AK002877.1 | - | 2, 3 | 150 | |||
| AK165424.1 | - | 2 | 172 | |||
| AK168495.1 | - | 2 | 124 | |||
| BC091743.1 | - | 2 | 109 | 5030475 | ||
| BY159585.1 | - | 2 | 220 | |||

Psma3
Genomic Map