siRNA Details
| Gene Symbol: Ptpro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164401.1 | NP_001157873.1 | 18 | 3125 | ||
| NM_001164402.1 | NP_001157874.1 | 8 | 755 | |||
| NM_001164403.1 | NP_001157875.1 | 7 | 671 | |||
| NM_011216.3 | NP_035346.3 | 19 | 3209 | |||
| GenBank mRNA | AF135166.1 | - | 18 | 3090 | ||
| AF295638.1 | - | 19 | 2822 | |||
| BC051976.1 | - | 8 | 668 | |||
| BC052743.1 | - | 19 | 3043 | 6834312 | ||
| BC063258.1 | - | 9 | 864 | |||
| BC079649.1 | - | 671 | ||||
| U37465.1 | - | 8 | 739 | |||
| U37466.1 | - | 7 | 655 | |||

Ptpro
Genomic Map