siRNA Details
| Gene Symbol: Sept4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284394.1 | NP_001271323.1 | 2 | 230 | ||
| NM_011129.2 | NP_035259.1 | 2 | 230 | |||
| GenBank mRNA | AK039859.1 | - | 3 | 240 | ||
| AK135143.1 | - | 2 | 231 | |||
| AK137299.1 | - | 2 | 149 | |||
| BC055101.1 | - | 2 | 230 | 6591016 | ||
| BU562316.1 | - | 3 | 132 | |||
| CJ144307.1 | - | 2 | 218 | |||
| X61452.1 | - | 2 | 160 | |||

Sept4
Genomic Map