siRNA Details
| Gene Symbol: STYX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130701.1 | NP_001124173.1 | 12 | 1499 | ||
| NM_145251.3 | NP_660294.1 | 11 | 1747 | |||
| XM_006720242.3 | XP_006720305.1 | 9 | 1367 | |||
| XM_011537108.2 | XP_011535410.1 | 11 | 1414 | |||
| XM_011537109.2 | XP_011535411.1 | 8 | 1308 | |||
| GenBank mRNA | AK098195.1 | - | 11 | 1729 | ||
| AL832591.1 | - | 12 | 1484 | |||
| AL832697.1 | - | 2 | 887 | |||
| BC020265.2 | - | 11 | 1698 | 3922829 | ||
| BC146995.1 | - | 11 | 1636 | 9056923 | ||
| BC146998.1 | - | 11 | 1637 | 9056926 | ||
| BX647636.1 | - | 11 | 1707 | |||

STYX
Genomic Map