siRNA Details
| Gene Symbol: Srgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011157.2 | NP_035287.1 | 2 | 202 | ||
| GenBank mRNA | AK087960.1 | - | 1 | 31 | ||
| AK151357.1 | - | 2 | 206 | |||
| AK157945.1 | - | 2 | 203 | |||
| AK168124.1 | - | 2 | 205 | |||
| BC037076.1 | - | 2 | 192 | 5038568 | ||
| J04549.1 | - | 2 | 199 | |||
| M34603.1 | - | 2 | 203 | |||
| X16133.1 | - | 2 | 202 | |||

Srgn
Genomic Map