siRNA Details
| Gene Symbol: Prl7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011168.4 | NP_035298.1 | 5 | 498 | ||
| GenBank mRNA | AF011382.1 | - | 5 | 487 | ||
| AF020524.1 | - | 5 | 494 | |||
| AK005455.1 | - | 5 | 499 | |||
| AK028068.1 | - | 5 | 498 | |||
| AK131665.1 | - | 5 | 499 | |||
| BC099468.1 | - | 5 | 477 | 30789902 | ||

Prl7a2
Genomic Map