siRNA Details
| Gene Symbol: Psmb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011185.3 | NP_035315.1 | 2 | 197 | ||
| GenBank mRNA | AK028265.1 | - | 2 | 219 | ||
| AK077520.1 | - | 2 | 198 | |||
| AK132161.1 | - | 2 | 198 | |||
| AK167647.1 | - | 2 | 219 | |||
| BC018351.1 | - | 2 | 198 | 3584538 | ||
| U60824.1 | - | 2 | 212 | |||
| X80686.1 | - | 2 | 201 | |||

Psmb1
Genomic Map