siRNA Details
| Gene Symbol: Psme1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011189.1 | NP_035319.1 | 4 | 263 | ||
| GenBank mRNA | AK008514.1 | - | 4 | 238 | ||
| AK019390.1 | - | 4 | 291 | |||
| AK150708.1 | - | 4 | 307 | |||
| AK150975.1 | - | 4 | 306 | |||
| BC088981.1 | - | 4 | 283 | 5689048 | ||
| D87909.1 | - | 4 | 263 | |||
| U60328.1 | - | 4 | 182 | |||

Psme1
Genomic Map